3-(4-Isobutylphenyl)acrylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Isobutylcinnamic acid

OR13177-1G OR13177-250MG
APOSOR13177-1GEA 24.8 EUR
APOSOR13177-1G APOSOR13177-250MG
3-(4-Isobutylphenyl)acrylic acid
3-(4-Isobutylphenyl)acrylic acid
Formel: C₁₃H₁₆O₂
Molecular Weight: 204.27 g/mol
Schmelzpunkt: 168…169 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00800640
CAS-Nummer: 66734-95-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR