Methyl-5-brom-2-benzo[b]thiophencarboxylat

Lieferant: Apollo Scientific

Synonyme: Methyl 5-bromobenzo[b]thiophene-2-carboxylate, Methyl 5-bromo-1-benzothiophene-2-carboxylate

OR13214-1G
APOSOR13214-1GEA 60.9 EUR
APOSOR13214-1G
Methyl-5-brom-2-benzo[b]thiophencarboxylat
Methyl-5-brom-2-benzo[b]thiophencarboxylat
Formel: C₁₀H₇BrO₂S
Molecular Weight: 271,1300 g/mol
Schmelzpunkt: 110…111 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05662034
CAS-Nummer: 7312-11-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR