3-Chloropyridine-4-boronic acid, neopentyl glycol ester

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(3-Chloropyridin-4-yl)-5,5-dimethyl-1,3,2-dioxaborinane, 3-Chloro-4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)pyridine

OR13217-250MG
APOSOR13217-250MGEA 81.2 EUR
APOSOR13217-250MG
3-Chloropyridine-4-boronic acid, neopentyl glycol ester
3-Chloropyridine-4-boronic acid, neopentyl glycol ester
Formel: C₁₀H₁₃BClNO₂
Molecular Weight: 225,48 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06796314
CAS-Nummer: 915070-52-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR