2-Amino-6-chlor-4-nitrophenol 97%
Lieferant: Apollo Scientific
Synonyme:
3-Chloro-2-hydroxy-5-nitroaniline
OR13236-25G
APOSOR13236-25GEA
139
EUR
APOSOR13236-25G
2-Amino-6-chlor-4-nitrophenol 97%
2-Amino-6-chlor-4-nitrophenol
Formel:
C₆H₅ClN₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00035767 CAS-Nummer: 6358-09-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...