3-Brom-5-nitro-2-pyridinol
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Bromo-5-nitro-2(1H)-pyridinone, 3-Bromo-5-nitro-2-pyridinol, 3-Bromo-5-nitro-2-pyridone
OR13237-1G
APOSOR13237-1GEA
21.9
EUR
APOSOR13237-1G
3-Brom-5-nitro-2-pyridinol
3-Bromo-2-hydroxy-5-nitropyridine
Formel:
C5H3BrN2O3 Molecular Weight: 218,99 g/mol Schmelzpunkt: 212…214 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03840431 CAS-Nummer: 15862-33-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...