3-Amino-3-(3-hydroxyphenyl)propanoic acid

Lieferant: Apollo Scientific
Achtung

OR13244-10G OR13244-1G OR13244-5G
APOSOR13244-10GEA 348 EUR
APOSOR13244-10G APOSOR13244-1G APOSOR13244-5G
3-Amino-3-(3-hydroxyphenyl)propanoic acid
3-Amino-3-(3-hydroxyphenyl)propanoic acid
Formel: C₉H₁₁NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01871309
CAS-Nummer: 102872-33-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR