2-Amino-N-tert-butylbenzamid

Lieferant: Apollo Scientific
Achtung

OR1326-1G
APOSOR1326-1GEA 79.9 EUR
APOSOR1326-1G
2-Amino-N-tert-butylbenzamid
2-Amino-N-tert-butylbenzamid
Formel: C₁₁H₁₆N₂O
Schmelzpunkt: 124…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00060601
CAS-Nummer: 1203-89-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR