(4-Methylsulphonylaminomethyl)benzeneboronic acid 98%

Lieferant: Apollo Scientific

OR1327-1G
APOSOR1327-1GEA 203 EUR
APOSOR1327-1G
(4-Methylsulphonylaminomethyl)benzeneboronic acid 98%
(4-Methylsulphonylaminomethyl)benzeneboronic acid
Formel: C₈H₁₂BNO₄S
Molecular Weight: 229,06 g/mol
Schmelzpunkt: 237…239 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659816
CAS-Nummer: 850568-38-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR