4-Amino-1H-pyrazole-5-carboxamide 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-1H-pyrazole-3-carboxamide

OR13272-250MG
APOSOR13272-250MGEA 119 EUR
APOSOR13272-250MG
4-Amino-1H-pyrazole-5-carboxamide 97%
4-Amino-1H-pyrazole-5-carboxamide
Formel: C₄H₆N₄O
Molecular Weight: 126,12 g/mol
Schmelzpunkt: 191 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02646740
CAS-Nummer: 67221-50-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR