1-(4-Amino-1,2,5-oxadiazol-3-yl)-5-methyl-1H-1,2,3-triazole-4-carboxamide

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-4-(4-carbamoyl-5-methyl-1H-1,2,3-triazol-1-yl)-1,2,5-oxadiazole, 1-(4-Amino-1,2,5-oxadiazol-3-yl)-4-carbamoyl-5-methyl-1H-1,2,3-triazole

OR13280-100MG
APOSOR13280-100MGEA 383 EUR
APOSOR13280-100MG
1-(4-Amino-1,2,5-oxadiazol-3-yl)-5-methyl-1H-1,2,3-triazole-4-carboxamide
1-(4-Amino-1,2,5-oxadiazol-3-yl)-5-methyl-1H-1,2,3-triazole-4-carboxamide
Formel: C₆H₇N₇O₂
Molecular Weight: 209,165 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01240850
CAS-Nummer: 312267-71-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR