Ethyl-4-amino-3-(aminocarbonyl)isothiazole-5-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 4-amino-3-carbamoylisothiazole-5-carboxylate
OR13281-1G
APOSOR13281-1GEA
371
EUR
APOSOR13281-1G
Ethyl-4-amino-3-(aminocarbonyl)isothiazole-5-carboxylate
Ethyl-4-amino-3-(aminocarbonyl)isothiazole-5-carboxylate
Formel:
C₇H₉N₃O₃S Molecular Weight: 215,23 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00848452 CAS-Nummer: 54968-74-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...