4-Hydroxy-3-nitrobenzeneboronic acid 98%
Lieferant: Apollo Scientific
Synonyme:
(4-Hydroxy-3-nitrophenyl)boronic acid
OR1330-1G
APOSOR1330-1GEA
174
EUR
APOSOR1330-1G
4-Hydroxy-3-nitrobenzeneboronic acid 98%
4-Hydroxy-3-nitrobenzeneboronic acid
Formel:
C₆H₆BNO₅ Molecular Weight: 182,927 g/mol Schmelzpunkt: 111…113 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06659843 CAS-Nummer: 850568-75-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...