2-Phenyl-1-(piperazin-1-yl)ethan-1-one

Lieferant: Apollo Scientific
Achtung

Synonyme: [2-Oxo-2-(piperazin-1-yl)ethyl]benzene, 1-(Phenylacetyl)piperazine

OR13308-100MG OR13308-250MG
APOSOR13308-100MGEA 125 EUR
APOSOR13308-100MG APOSOR13308-250MG
2-Phenyl-1-(piperazin-1-yl)ethan-1-one
2-Phenyl-1-(piperazin-1-yl)ethan-1-one
Formel: C₁₂H₁₆N₂O
Siedepunkt: 190…191 °C (17 mm Hg)
Lagertemperatur: Kühlschrank
MDL: MFCD01566912
CAS-Nummer: 88372-33-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR