3-Amino-3-(3-methoxyphenyl)propionsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Amino-3-(3-methoxyphenyl)propanoic acid
OR13354-1G
APOSOR13354-1GEA
21.9
EUR
APOSOR13354-1G
3-Amino-3-(3-methoxyphenyl)propionsäure
3-Amino-3-(3-methoxyphenyl)propionsäure
Formel:
C₁₀H₁₃NO₃ Lagertemperatur: Kühlschrank |
MDL:
MFCD00462569 CAS-Nummer: 68208-19-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...