3-Amino-4-chlorphenylboronsäurepinakolester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-4-chlorobenzeneboronic acid pinacol ester, 3-Amino-4-chlorophenylboronic acid, pinacol ester, 2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

OR1336-1G OR1336-250MG OR1336-5G
APOSOR1336-1GEA 183 EUR
APOSOR1336-1G APOSOR1336-250MG APOSOR1336-5G
3-Amino-4-chlorphenylboronsäurepinakolester 98%
3-Amino-4-chlorphenylboronsäurepinakolester
Formel: C₁₂H₁₇BClNO₂
Schmelzpunkt: 142…144 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659926
CAS-Nummer: 850567-56-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR