5-Amino-2-phenyl-2H-1,2,3-triazole-4-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-4-carboxy-2-phenyl-2H-1,2,3-triazole

OR13370-250MG
APOSOR13370-250MGEA 348 EUR
APOSOR13370-250MG
5-Amino-2-phenyl-2H-1,2,3-triazole-4-carboxylic acid
5-Amino-2-phenyl-2H-1,2,3-triazole-4-carboxylic acid
Formel: C₉H₈N₄O₂
Molecular Weight: 204,185 g/mol
Schmelzpunkt: 239 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02641259
CAS-Nummer: 400073-84-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR