Ethyl-2-amino-4-oxazolcarboxylat

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-aminooxazole-4-carboxylate

OR1342-1G
APOSOR1342-1GEA 21.9 EUR
APOSOR1342-1G
Ethyl-2-amino-4-oxazolcarboxylat
Ethyl-2-amino-4-oxazolcarboxylat
Formel: C₆H₈N₂O₃
Molecular Weight: 156,14 g/mol
Schmelzpunkt: 124…128 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06659907
CAS-Nummer: 177760-52-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR