4-Amino-3-nitrobenzoesäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Amino-3-nitrobenzoic Acid
OR13440-25G
APOSOR13440-25GEA
21.9
EUR
APOSOR13440-25G
4-Amino-3-nitrobenzoesäure 95%
4-Amino-3-nitrobenzoic acid
Formel:
C₇H₆N₂O₄ Molecular Weight: 182,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017009 CAS-Nummer: 1588-83-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...