1-Methyl-4-nitro-1H-pyrazole-5-carboxylic acid

Lieferant: Apollo Scientific

OR13452-1G
APOSOR13452-1GEA 77 EUR
APOSOR13452-1G
1-Methyl-4-nitro-1H-pyrazole-5-carboxylic acid
1-Methyl-4-nitro-1H-pyrazole-5-carboxylic acid
Formel: C₅H₅N₃O₄
Molecular Weight: 171.111 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00222738
CAS-Nummer: 92534-69-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR