4-Amino-3-phenylbutanoic acid hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
β-(Carboxymethyl)phenethylamine hydrochloride
OR13459-1G
APOSOR13459-1GEA
21.9
EUR
APOSOR13459-1G
4-Amino-3-phenylbutanoic acid hydrochloride
4-Amino-3-phenylbutanoic acid hydrochloride
Formel:
C₁₀H₁₃NO₂·ClH Molecular Weight: 215,677 g/mol Schmelzpunkt: 194…201 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075860 CAS-Nummer: 3060-41-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...