6-Methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-sulphonyl chloride 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 6-Methyluracil-5-sulphonyl chloride

OR13481-1G
APOSOR13481-1GEA 516 EUR
APOSOR13481-1G
6-Methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-sulphonyl chloride 95%
6-Methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-sulphonyl chloride
Formel: C₅H₅ClN₂O₄S
Molecular Weight: 224.62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00476725
CAS-Nummer: 6461-30-9
UN: 1759
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR