1H-Imidazo[4,5-b]pyridine-2-thiol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1H-Imidazo[4,5-b]pyridine-2(3H)-thione, 2-Thio-1H-imidazo[4,5-b]pyridine
OR13527-250MG
APOSOR13527-250MGEA
21.9
EUR
APOSOR13527-250MG
1H-Imidazo[4,5-b]pyridine-2-thiol
1H-Imidazo[4,5-b]pyridine-2-thiol
Formel:
C₆H₅N₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00022663 CAS-Nummer: 29448-81-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...