4-tert-Butylbenzolsulfonylchlorid
Lieferant: Apollo Scientific
|
Gefahr
|
OR1355-25G
OR1355-5G
APOSOR1355-25GEA
50.9
EUR
APOSOR1355-25G
APOSOR1355-5G
4-tert-Butylbenzolsulfonylchlorid
4-tert-Butylbenzolsulfonylchlorid
Formel:
C₁₀H₁₃ClO₂S Schmelzpunkt: 78…81 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007449 CAS-Nummer: 15084-51-2 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...