7-Nitro-1H-indazol 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
7-NINA, 7-Nitro-1H-indazol
OR13550-25G
OR13550-5G
APOSOR13550-25GEA
102
EUR
APOSOR13550-25G
APOSOR13550-5G
7-Nitro-1H-indazol 97%
7-Nitro-1H-indazole
Formel:
C₇H₅N₃O₂ Molecular Weight: 163.14 g/mol Schmelzpunkt: 190 Lagertemperatur: Gefrierschrank |
MDL:
MFCD00022789 CAS-Nummer: 2942-42-9 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...