2-Nitro-1-phenyl-1-ethanol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Nitro-1-phenylethanol

OR13560-1G
APOSOR13560-1GEA 810 EUR
APOSOR13560-1G
2-Nitro-1-phenyl-1-ethanol
2-Nitro-1-phenyl-1-ethanol
Formel: C₈H₉NO₃
Molecular Weight: 167,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00854219
CAS-Nummer: 15990-45-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR