AKR1C3 Inhibitor I 98%

Lieferant: Apollo Scientific

Synonyme: 3-(3,4-Dihydroisoquinolin-2(1H)-ylsulphonyl)benzoic acid, AKR1C3-IN-1, 3-((3,4-Dihydroisoquinolin-2(1H)-yl)sulphonyl)benzoic acid

OR13578-1G
APOSOR13578-1GEA 128 EUR
APOSOR13578-1G
AKR1C3 Inhibitor I 98%
AKR1C3 Inhibitor I
Formel: C₁₆H₁₅NO₄S
Molecular Weight: 317,36 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01197550
CAS-Nummer: 327092-81-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR