Methyl-p-tolylsulfon 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-(Methylsulphonyl)toluene, 1-Methyl-4-(methylsulphonyl)benzene

OR1358-25G
APOSOR1358-25GEA 24.8 EUR
APOSOR1358-25G
Methyl-p-tolylsulfon 98%
Methyl-p-tolylsulfon
Formel: C₈H₁₀O₂S
Molecular Weight: 170,23 g/mol
Schmelzpunkt: 85…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014742
CAS-Nummer: 3185-99-7
EINECS: 221-682-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR