2,6-Dibrom-3-nitropyridin

Lieferant: Apollo Scientific
Achtung

OR13599-5G
APOSOR13599-5GEA 27.7 EUR
APOSOR13599-5G
2,6-Dibrom-3-nitropyridin
2,6-Dibromo-3-nitropyridine
Formel: C₅H₂Br₂N₂O₂
Molecular Weight: 281,8900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00233995
CAS-Nummer: 55304-80-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR