2-Cyan-6-methylpyridin 97%
Lieferant: Apollo Scientific
Synonyme:
6-Methyl-2-pyridincarbonitril, 6-Cyan-2-picolin
OR13606-100G
APOSOR13606-100GEA
159
EUR
APOSOR13606-100G
2-Cyan-6-methylpyridin 97%
2-Cyano-6-methylpyridine
Formel:
C₇H₆N₂ Molecular Weight: 118.14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00190586 CAS-Nummer: 1620-75-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...