3-Chlor-2-hydroxypyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloro-2-pyridinol

OR13614-1G OR13614-5G OR13614-25G
APOSOR13614-1GEA 21.9 EUR
APOSOR13614-1G APOSOR13614-5G APOSOR13614-25G
3-Chlor-2-hydroxypyridin
3-Chlor-2-hydroxypyridin
Formel: C₅H₄ClNO
Molecular Weight: 129.55 g/mol
Schmelzpunkt: 177 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01646109
CAS-Nummer: 13466-35-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR