3-Chlor-2-hydroxypyridin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Chloro-2-pyridinol
OR13614-1G
OR13614-5G
OR13614-25G
APOSOR13614-1GEA
21.9
EUR
APOSOR13614-1G
APOSOR13614-5G
APOSOR13614-25G
3-Chlor-2-hydroxypyridin
3-Chlor-2-hydroxypyridin
Formel:
C₅H₄ClNO Molecular Weight: 129.55 g/mol Schmelzpunkt: 177 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01646109 CAS-Nummer: 13466-35-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...