4-Hydroxy-3-pyridinsulfonsäure 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR13620-25G
OR13620-5G
OR13620-10G
APOSOR13620-25GEA
0
EUR
APOSOR13620-25G
APOSOR13620-5G
APOSOR13620-10G
4-Hydroxy-3-pyridinsulfonsäure 97%
4-Hydroxy-3-pyridinesulphonic acid
Formel:
C₅H₅NO₄S Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093473 CAS-Nummer: 51498-37-4 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...