Pyridine hydrobromide 97%
Lieferant: Apollo Scientific
Synonyme:
Pyridinium Bromide
OR13635-500G
APOSOR13635-500GEA
33.5
EUR
APOSOR13635-500G
Pyridine hydrobromide 97%
Pyridine hydrobromide
Formel:
C₅H₅N Molecular Weight: 160,01 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012801 CAS-Nummer: 18820-82-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...