1-(2,4-Dichlorophenyl)-2-(1-imidazolyl)ethanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-[2-(2,4-Dichlorophenyl)-2-hydroxyethyl]imidazole, 1-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-yl)ethanol
OR13643-100G
APOSOR13643-100GEA
27.7
EUR
APOSOR13643-100G
1-(2,4-Dichlorophenyl)-2-(1-imidazolyl)ethanol
1-(2,4-Dichlorophenyl)-2-(1-imidazolyl)ethanol
Formel:
C₁₁H₁₀Cl₂N₂O Molecular Weight: 257,11 g/mol Schmelzpunkt: 135 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00044708 CAS-Nummer: 24155-42-8 EINECS: 246-042-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...