Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate, Methyl 3,4-dihydro-3-oxo-2H-1,4-benzothiazine-6-carboxylate

OR13646-1G
APOSOR13646-1GEA 21.9 EUR
APOSOR13646-1G
Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate
Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate
Formel: C₁₀H₉NO₃S
Molecular Weight: 223,25 g/mol
Schmelzpunkt: 97…99 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00449104
CAS-Nummer: 188614-01-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR