5-Chloro-3-(2-hydroxyethyl)-1H-indole 95%

Lieferant: Apollo Scientific
Achtung

OR13671-250MG
APOSOR13671-250MGEA 88.6 EUR
APOSOR13671-250MG
5-Chloro-3-(2-hydroxyethyl)-1H-indole 95%
5-Chloro-3-(2-hydroxyethyl)-1H-indole
Formel: C₁₀H₁₀ClNO
Lagertemperatur: Kühlschrank
MDL: MFCD00130166
CAS-Nummer: 61220-51-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR