3-Amino-5-ethoxypyridine 98%

Lieferant: Apollo Scientific

Synonyme: 5-Ethoxypyridin-3-amine

OR13672-250MG
APOSOR13672-250MGEA 79.9 EUR
APOSOR13672-250MG
3-Amino-5-ethoxypyridine 98%
3-Amino-5-ethoxypyridine
Formel: C₇H₁₀N₂O
Molecular Weight: 138,167 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01646192
CAS-Nummer: 51468-00-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR