2-(3-Ethoxy-4-ethoxycarbonylphenyl)acetic acid
Lieferant: Apollo Scientific
|
Gefahr
|
OR13687-25G
APOSOR13687-25GEA
68.3
EUR
APOSOR13687-25G
2-(3-Ethoxy-4-ethoxycarbonylphenyl)acetic acid
2-(3-Ethoxy-4-ethoxycarbonylphenyl)acetic acid
Formel:
C₁₃H₁₆O₅ Schmelzpunkt: 80 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08704233 CAS-Nummer: 99469-99-5 EINECS: 427-630-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...