3-Pyridin-4-ylpropionic acid 98%

Lieferant: Apollo Scientific
Achtung

OR1370-5G
APOSOR1370-5GEA 140 EUR
APOSOR1370-5G
3-Pyridin-4-ylpropionic acid 98%
3-Pyridin-4-ylpropionic acid
Formel: C₈H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00084846
CAS-Nummer: 6318-43-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR