6-Methylnicotinsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Methyl-3-pyridincarbonsäure, 6-Methylpyridin-3-carbonsäure

OR13706-25G OR13706-1G
APOSOR13706-25GEA 65.4 EUR
APOSOR13706-25G APOSOR13706-1G
6-Methylnicotinsäure 98%
6-Methylnicotinsäure
Formel: C₇H₇NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00006341
CAS-Nummer: 3222-47-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR