3-Isopropylbenzoic acid 98%

Lieferant: Apollo Scientific
Achtung

OR13714-5G
APOSOR13714-5GEA 120 EUR
APOSOR13714-5G
3-Isopropylbenzoic acid 98%
3-Isopropylbenzoic acid
Formel: C10H12O2
Molecular Weight: 164.2011 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09038906
CAS-Nummer: 5651-47-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR