3,5-Dibrom-2-methylthiophen

Lieferant: Apollo Scientific
Achtung

OR13720-1G
APOSOR13720-1GEA 21.9 EUR
APOSOR13720-1G
3,5-Dibrom-2-methylthiophen
3,5-Dibromo-2-methylthiophene
Formel: C₅H₄Br₂S
Molecular Weight: 255,9583 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07781197
CAS-Nummer: 29421-73-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR