4-Bromanthranilsäure 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Amino-4-bromobenzoic acid

OR13732-100G OR13732-25G OR13732-5G
APOSOR13732-100GEA 66.7 EUR
APOSOR13732-100G APOSOR13732-25G APOSOR13732-5G
4-Bromanthranilsäure 95%
4-Bromanthranilsäure
Formel: C₇H₆BrNO₂
Molecular Weight: 216.03 g/mol
Schmelzpunkt: 230…234 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03618454
CAS-Nummer: 20776-50-5
EINECS: 244-025-7
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR