Methyl 4-chloro-3-(cyanomethyl)benzoate 98%

Lieferant: Apollo Scientific
Achtung

OR13734-5G
APOSOR13734-5GEA 0 EUR
APOSOR13734-5G
Methyl 4-chloro-3-(cyanomethyl)benzoate 98%
Methyl 4-chloro-3-(cyanomethyl)benzoate
Formel: C₁₀H₈ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09263465
CAS-Nummer: 872091-83-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR