2-Amino-5-(4-methylphenyl)-1,3,4-thiadiazole 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(5-Amino-1,3,4-thiadiazol-2-yl)toluene, 5-(4-Methylphenyl)-1,3,4-thiadiazol-2-amine, 5-(p-Tolyl)-1,3,4-thiadiazol-2-amine

OR13741-5G
APOSOR13741-5GEA 184 EUR
APOSOR13741-5G
2-Amino-5-(4-methylphenyl)-1,3,4-thiadiazole 95+%
2-Amino-5-(4-methylphenyl)-1,3,4-thiadiazole
Formel: C₉H₉N₃S
Molecular Weight: 191,253 g/mol
Schmelzpunkt: 218…222 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00469688
CAS-Nummer: 26907-54-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR