5-Phenyl-1,3,4-oxadiazole-2-thiol 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Phenyl-1,3,4-oxadiazole-2(3H)-thione
OR13743-25G
APOSOR13743-25GEA
68.3
EUR
APOSOR13743-25G
5-Phenyl-1,3,4-oxadiazole-2-thiol 95%
5-Phenyl-1,3,4-oxadiazole-2-thiol
Formel:
C₈H₆N₂OS Molecular Weight: 178,21 g/mol Schmelzpunkt: 219…222 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00030467 CAS-Nummer: 3004-42-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...