7-Amino-2H-1,4-benzoxazin-3(4H)-one

Lieferant: Apollo Scientific

Synonyme: 7-Amino-2H-1,4-benzoxazin-3[4H]one

OR13747-1G
APOSOR13747-1GEA 0 EUR
APOSOR13747-1G
7-Amino-2H-1,4-benzoxazin-3(4H)-one
7-Amino-2H-1,4-benzoxazin-3(4H)-one
Formel: C₈H₈N₂O₂
Molecular Weight: 164,17 g/mol
Schmelzpunkt: 220…222 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03425794
CAS-Nummer: 26215-14-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR