8-Amino-4H-1,4-benzoxazin-3-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 8-Amino-2H-1,4-benzoxazin-3(4H)-one

OR13819-250MG
APOSOR13819-250MGEA 72.5 EUR
APOSOR13819-250MG
8-Amino-4H-1,4-benzoxazin-3-one
8-Amino-4H-1,4-benzoxazin-3-one
Formel: C₈H₈N₂O₂
Molecular Weight: 164,17 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09998373
CAS-Nummer: 321126-82-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR