tert-Butyl-4-(3-aminobenzyl)piperazine-1-carboxylate 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR1382-250MG
APOSOR1382-250MGEA
21.9
EUR
APOSOR1382-250MG
tert-Butyl-4-(3-aminobenzyl)piperazine-1-carboxylate 97%
tert-Butyl-4-(3-aminobenzyl)piperazine-1-carboxylate
Formel:
C₁₆H₂₅N₃O₂ Schmelzpunkt: 107…108 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06658979 CAS-Nummer: 361345-40-6 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...