Methyl-5-brom-6-chlornicotinat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 5-bromo-6-chloronicotinate

OR13854-25G OR13854-1G OR13854-5G
APOSOR13854-25GEA 116 EUR
APOSOR13854-25G APOSOR13854-1G APOSOR13854-5G
Methyl-5-brom-6-chlornicotinat
Methyl-5-brom-6-chlornicotinat
Formel: C₇H₅BrClNO₂
Schmelzpunkt: 76…79 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03844855
CAS-Nummer: 78686-77-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR